Assay Details
| Gene Symbol: LOC4328053 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769282.2 | XP_015624768.1 | 7 - 8 | 582 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4328053| Gene Symbol: LOC4328053 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769282.2 | XP_015624768.1 | 7 - 8 | 582 | 90 | |
Set Membership: |
|