Assay Details
| Gene Symbol: LOC4328183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767273.2 | XP_015622759.1 | 8 - 9 | 1839 | 101 | |
| GenBank mRNA | BT131737.1 | - | 1 - 2 | 221 | 101 | |
More Information
Set Membership: |
|

LOC4328183
Genomic Map