Assay Details
| Gene Symbol: LOC4328183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767273.2 | XP_015622759.1 | 9 - 10 | 2060 | 130 | |
| GenBank mRNA | AK120049.1 | - | 9 - 10 | 2057 | 130 | |
| BT131737.1 | - | 2 - 3 | 442 | 130 | ||
More Information
Set Membership: |
|

LOC4328183
Genomic Map