Assay Details
| Gene Symbol: LOC4328430 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770853.2 | XP_015626339.1 | 3 - 4 | 433 | 148 | |
| XM_015770854.2 | XP_015626340.1 | 3 - 4 | 460 | 148 | ||
| XM_015770855.2 | XP_015626341.1 | 3 - 4 | 440 | 148 | ||
| XM_015770856.2 | XP_015626342.1 | 3 - 4 | 428 | 148 | ||
More Information
Set Membership: |
|

LOC4328430
Genomic Map