Assay Details
| Gene Symbol: LOC4328430 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770853.2 | XP_015626339.1 | 9 - 10 | 818 | 102 | |
| XM_015770854.2 | XP_015626340.1 | 9 - 10 | 845 | 102 | ||
| XM_015770855.2 | XP_015626341.1 | 9 - 10 | 825 | 102 | ||
| XM_015770856.2 | XP_015626342.1 | 9 - 10 | 813 | 102 | ||
| GenBank mRNA | AK067291.1 | - | 9 - 10 | 814 | 102 | |
More Information
Set Membership: |
|

LOC4328430
Genomic Map