Assay Details
| Gene Symbol: LOC4328745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771615.2 | XP_015627101.1 | 16 - 17 | 2478 | 64 | |
| GenBank mRNA | AK059059.1 | - | 11 - 12 | 1388 | 64 | |
| AK059794.1 | - | 5 - 6 | 734 | 64 | ||
| AK061043.1 | - | 3 - 4 | 477 | 64 | ||
More Information
Set Membership: |
|

LOC4328745
Genomic Map