Assay Details
| Gene Symbol: LOC4328745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771615.2 | XP_015627101.1 | 7 - 8 | 1205 | 105 | |
| GenBank mRNA | AK059059.1 | - | 2 - 3 | 115 | 105 | |
More Information
Set Membership: |
|

LOC4328745
Genomic Map