Assay Details
| Gene Symbol: LOC4328916 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771556.2 | XP_015627042.1 | 1 - 2 | 358 | 62 | |
| XM_015771557.1 | XP_015627043.1 | 1 - 2 | 373 | 62 | ||
| GenBank mRNA | AK059303.1 | - | 1 - 2 | 342 | 62 | |
| AK064805.1 | - | 1 - 2 | 351 | 62 | ||
| AK104581.1 | - | 1 - 2 | 341 | 62 | ||
More Information
Set Membership: |
|

LOC4328916
Genomic Map