Assay Details
| Gene Symbol: LOC4328916 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771556.2 | XP_015627042.1 | 2 - 3 | 571 | 82 | |
| GenBank mRNA | AK059303.1 | - | 2 - 3 | 555 | 82 | |
| AK064805.1 | - | 2 - 3 | 564 | 82 | ||
| AK104581.1 | - | 2 - 3 | 554 | 82 | ||
More Information
Set Membership: |
|

LOC4328916
Genomic Map