Assay Details
| Gene Symbol: LOC4329037 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771915.2 | 2 - 3 | 351 | 102 | ||
| GenBank mRNA | AK072177.1 | - | 2 - 3 | 356 | 102 | |
| AK099035.1 | - | 2 - 3 | 352 | 102 | ||
More Information
Set Membership: |
|

LOC4329037
Genomic Map