Assay Details
| Gene Symbol: LOC4329037 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771915.2 | 8 - 9 | 1222 | 89 | ||
| GenBank mRNA | AK060415.1 | - | 5 - 6 | 539 | 89 | |
| AK072177.1 | - | 8 - 9 | 1227 | 89 | ||
| AK099035.1 | - | 8 - 9 | 1222 | 89 | ||
More Information
Set Membership: |
|

LOC4329037
Genomic Map