Assay Details
| Gene Symbol: LOC4329037 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771915.2 | 9 - 10 | 1383 | 104 | ||
| GenBank mRNA | AK060415.1 | - | 6 - 7 | 700 | 104 | |
| AK072177.1 | - | 9 - 10 | 1388 | 104 | ||
| AK099035.1 | - | 9 - 10 | 1383 | 104 | ||
More Information
Set Membership: |
|

LOC4329037
Genomic Map