Assay Details
| Gene Symbol: LOC4329067 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771314.2 | XP_015626800.1 | 12 - 13 | 1434 | 129 | |
| GenBank mRNA | AK242797.1 | - | 3 - 4 | 224 | 129 | |
More Information
Set Membership: |
|

LOC4329067
Genomic Map