Assay Details
| Gene Symbol: LOC4329067 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771314.2 | XP_015626800.1 | 13 - 14 | 1549 | 96 | |
| GenBank mRNA | AK242797.1 | - | 4 - 5 | 339 | 96 | |
More Information
Set Membership: |
|

LOC4329067
Genomic Map