Assay Details
| Gene Symbol: LOC4329067 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771314.2 | XP_015626800.1 | 14 - 15 | 1711 | 93 | |
| GenBank mRNA | AK242797.1 | - | 5 - 6 | 501 | 93 | |
More Information
Set Membership: |
|

LOC4329067
Genomic Map