Assay Details
| Gene Symbol: LOC4329134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK102001.1 | - | 14 - 15 | 1393 | 130 | |
| AK112066.1 | - | 14 - 15 | 1390 | 130 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4329134| Gene Symbol: LOC4329134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK102001.1 | - | 14 - 15 | 1393 | 130 | |
| AK112066.1 | - | 14 - 15 | 1390 | 130 | ||
Set Membership: |
|