Assay Details
| Gene Symbol: LOC112936002 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771663.2 | XP_015627149.1 | 7 - 8 | 1008 | 85 | |
| XM_015771664.2 | XP_015627150.1 | 8 - 9 | 913 | 85 | ||
| XM_015771665.2 | XP_015627151.1 | 7 - 8 | 1625 | 85 | ||
| XM_015771666.2 | XP_015627152.1 | 7 - 8 | 998 | 85 | ||
| XM_015771667.2 | XP_015627153.1 | 7 - 8 | 1613 | 85 | ||
| GenBank mRNA | AK070515.1 | - | 7 - 8 | 908 | 85 | |
More Information
Set Membership: |
|

LOC112936002
Genomic Map