Assay Details
| Gene Symbol: LOC4329546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767708.2 | XP_015623194.1 | 17 - 18 | 2712 | 73 | |
| GenBank mRNA | AK102681.1 | - | 17 - 18 | 2482 | 73 | |
| AK105277.1 | - | 5 - 6 | 674 | 73 | ||
More Information
Set Membership: |
|

LOC4329546
Genomic Map