Assay Details
| Gene Symbol: LOC4329678 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771102.2 | XP_015626588.1 | 7 - 8 | 2681 | 97 | |
| XM_015771103.2 | XP_015626589.1 | 8 - 9 | 2724 | 97 | ||
| XM_015771104.2 | XP_015626590.1 | 8 - 9 | 2714 | 97 | ||
| XM_015771105.2 | XP_015626591.1 | 7 - 8 | 2676 | 97 | ||
| XM_015771106.2 | XP_015626592.1 | 7 - 8 | 2676 | 97 | ||
| XM_015771107.2 | XP_015626593.1 | 7 - 8 | 2559 | 97 | ||
| GenBank mRNA | AB069967.1 | - | 6 - 7 | 2163 | 97 | |
| AK062032.1 | - | 1 - 2 | 982 | 97 | ||
| AK066353.1 | - | 7 - 8 | 2701 | 97 | ||
| AK069669.1 | - | 7 - 8 | 2551 | 97 | ||
More Information
Set Membership: |
|

LOC4329678
Genomic Map