Assay Details
| Gene Symbol: LOC4329738 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769915.2 | XP_015625401.1 | 1 - 2 | 826 | 76 | |
| XM_015769916.2 | XP_015625402.1 | 1 - 2 | 824 | 76 | ||
| GenBank mRNA | AB024337.1 | - | 1 - 2 | 693 | 76 | |
| AB073546.1 | - | 1 - 2 | 419 | 76 | ||
| AK064910.1 | - | 1 - 2 | 744 | 76 | ||
More Information
Set Membership: |
|

LOC4329738
Genomic Map