Assay Details
| Gene Symbol: LOC4329744 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767277.2 | XP_015622763.1 | 2 - 3 | 469 | 79 | |
| XM_015767278.2 | XP_015622764.1 | 2 - 3 | 477 | 79 | ||
| GenBank mRNA | AK065365.1 | - | 2 - 3 | 311 | 79 | |
| AK103980.1 | - | 2 - 3 | 208 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015767277.2, XM_015767278.2 transcripts.
Set Membership: |
|

LOC4329744
Genomic Map