Assay Details
| Gene Symbol: LOC4329751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768992.2 | XP_015624478.1 | 11 - 12 | 1759 | 94 | |
| GenBank mRNA | AK058407.1 | - | 5 - 6 | 415 | 94 | |
| AK099974.1 | - | 11 - 12 | 1595 | 94 | ||
More Information
Set Membership: |
|

LOC4329751
Genomic Map