Assay Details
| Gene Symbol: LOC4329751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768992.2 | XP_015624478.1 | 19 - 20 | 2420 | 62 | |
| GenBank mRNA | AK058407.1 | - | 13 - 14 | 1076 | 62 | |
| AK099974.1 | - | 19 - 20 | 2256 | 62 | ||
More Information
Set Membership: |
|

LOC4329751
Genomic Map