Assay Details
| Gene Symbol: LOC4329751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768992.2 | XP_015624478.1 | 7 - 8 | 1465 | 70 | |
| GenBank mRNA | AK058407.1 | - | 1 - 2 | 121 | 70 | |
| AK099974.1 | - | 7 - 8 | 1301 | 70 | ||
More Information
Set Membership: |
|

LOC4329751
Genomic Map