Assay Details
| Gene Symbol: LOC4329908 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767888.2 | XP_015623374.1 | 1 | 1883 | 93 | |
| GenBank mRNA | AK287597.1 | - | 1 | 1855 | 93 | |
More Information
Set Membership: |
|

LOC4329908
Genomic Map