Assay Details
| Gene Symbol: LOC4329963 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769822.2 | XP_015625308.1 | 7 - 8 | 879 | 110 | |
| GenBank mRNA | AK061262.1 | - | 7 - 8 | 832 | 110 | |
| AK069360.1 | - | 7 - 8 | 822 | 110 | ||
| AK101546.1 | - | 7 - 8 | 833 | 110 | ||
| AK288426.1 | - | 2610 | 110 | |||
| AY278997.1 | - | 7 - 8 | 650 | 110 | ||
More Information
Set Membership: |
|

LOC4329963
Genomic Map