Assay Details
| Gene Symbol: LOC4330479 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770349.2 | XP_015625835.1 | 6 - 7 | 4543 | 93 | |
| XM_015770350.2 | XP_015625836.1 | 7 - 8 | 4414 | 93 | ||
| GenBank mRNA | AK108743.1 | - | 1 - 2 | 977 | 93 | |
More Information
Set Membership: |
|

LOC4330479
Genomic Map