Assay Details
| Gene Symbol: LOC4330696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768060.2 | XP_015623546.1 | 22 - 23 | 2577 | 81 | |
| GenBank mRNA | AK106279.1 | - | 4 - 5 | 241 | 81 | |
More Information
Set Membership: |
|

LOC4330696
Genomic Map