Assay Details
| Gene Symbol: LOC4330696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768060.2 | XP_015623546.1 | 24 - 25 | 2983 | 126 | |
| GenBank mRNA | AK106279.1 | - | 6 - 7 | 647 | 126 | |
More Information
Set Membership: |
|

LOC4330696
Genomic Map