Assay Details
| Gene Symbol: LOC4330747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767928.2 | XP_015623414.1 | 4 - 5 | 865 | 114 | |
| GenBank mRNA | AK101802.1 | - | 4 - 5 | 749 | 114 | |
More Information
Set Membership: |
|

LOC4330747
Genomic Map