Assay Details
| Gene Symbol: LOC4331508 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774258.2 | XP_015629744.1 | 3 - 4 | 1258 | 98 | |
| XM_015774259.2 | XP_015629745.1 | 2 - 3 | 1238 | 98 | ||
| GenBank mRNA | AK100046.1 | - | 2 - 3 | 1002 | 98 | |
More Information
Set Membership: |
|

LOC4331508
Genomic Map