Assay Details
| Gene Symbol: LOC4331547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775923.2 | XP_015631409.1 | 18 - 19 | 2667 | 99 | |
| GenBank mRNA | AK061677.1 | - | 3 - 4 | 615 | 99 | |
| AK067183.1 | - | 18 - 19 | 2674 | 99 | ||
More Information
Set Membership: |
|

LOC4331547
Genomic Map