Assay Details
| Gene Symbol: LOC4331547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775923.2 | XP_015631409.1 | 7 - 8 | 1140 | 72 | |
| GenBank mRNA | AK067183.1 | - | 7 - 8 | 1147 | 72 | |
More Information
Set Membership: |
|

LOC4331547
Genomic Map