Assay Details
| Gene Symbol: LOC4331556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774893.2 | XP_015630379.1 | 9 - 10 | 2459 | 98 | |
| XM_015774894.2 | XP_015630380.1 | 9 - 10 | 2451 | 98 | ||
| XM_015774895.2 | XP_015630381.1 | 10 - 11 | 2660 | 98 | ||
| GenBank mRNA | AK066407.1 | - | 10 - 11 | 2635 | 98 | |
More Information
Set Membership: |
|

LOC4331556
Genomic Map