Assay Details
| Gene Symbol: LOC4331611 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777732.1 | XP_015633218.1 | 5 - 6 | 1473 | 67 | |
| GenBank mRNA | AK111974.1 | - | 5 - 6 | 1373 | 67 | |
| AK120554.1 | - | 5 - 6 | 1330 | 67 | ||
More Information
Set Membership: |
|

LOC4331611
Genomic Map