Assay Details
| Gene Symbol: LOC4332540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774680.2 | XP_015630166.1 | 1 | 607 | 124 | |
| GenBank mRNA | AK109176.1 | - | 1 | 256 | 124 | |
More Information
Set Membership: |
|

LOC4332540
Genomic Map