Assay Details
| Gene Symbol: LOC4332700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772956.2 | XP_015628442.1 | 2 - 3 | 276 | 88 | |
| GenBank mRNA | AK059868.1 | - | 2 - 3 | 262 | 88 | |
| AK071431.1 | - | 2 - 3 | 258 | 88 | ||
More Information
Set Membership: |
|

LOC4332700
Genomic Map