Assay Details
| Gene Symbol: LOC4332704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772298.1 | XP_015627784.1 | 4 - 5 | 534 | 82 | |
| XM_015772299.1 | XP_015627785.1 | 4 - 5 | 525 | 82 | ||
| GenBank mRNA | AK100282.1 | - | 4 - 5 | 525 | 82 | |
More Information
Set Membership: |
|

LOC4332704
Genomic Map