Assay Details
| Gene Symbol: LOC4332782 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773716.1 | XP_015629202.1 | 2 - 3 | 495 | 137 | |
| GenBank mRNA | AK061545.1 | - | 2 - 3 | 495 | 137 | |
| AK067107.1 | - | 2 - 3 | 491 | 137 | ||
| AK104156.1 | - | 2 - 3 | 492 | 137 | ||
| AK104395.1 | - | 2 - 3 | 495 | 137 | ||
| AK104470.1 | - | 2 - 3 | 495 | 137 | ||
| AK104487.1 | - | 2 - 3 | 495 | 137 | ||
| AK104494.1 | - | 2 - 3 | 495 | 137 | ||
| AK104508.1 | - | 2 - 3 | 495 | 137 | ||
| AK104573.1 | - | 2 - 3 | 495 | 137 | ||
| AK104644.1 | - | 2 - 3 | 495 | 137 | ||
| AK106109.1 | - | 2 - 3 | 495 | 137 | ||
More Information
Set Membership: |
|

LOC4332782
Genomic Map