Assay Details
| Gene Symbol: LOC4332782 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773716.1 | XP_015629202.1 | 3 - 4 | 670 | 70 | |
| GenBank mRNA | AK067107.1 | - | 3 - 4 | 666 | 70 | |
| AK104156.1 | - | 3 - 4 | 667 | 70 | ||
| AK104395.1 | - | 3 - 4 | 670 | 70 | ||
| AK104470.1 | - | 3 - 4 | 670 | 70 | ||
| AK104487.1 | - | 3 - 4 | 670 | 70 | ||
| AK104494.1 | - | 3 - 4 | 670 | 70 | ||
| AK104508.1 | - | 3 - 4 | 670 | 70 | ||
| AK104573.1 | - | 3 - 4 | 670 | 70 | ||
| AK104644.1 | - | 3 - 4 | 670 | 70 | ||
| AK106109.1 | - | 3 - 4 | 670 | 70 | ||
More Information
Set Membership: |
|

LOC4332782
Genomic Map