Assay Details
| Gene Symbol: LOC9269259 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777817.2 | XP_015633303.1 | 1 - 2 | 374 | 103 | |
| GenBank mRNA | AK059731.1 | - | 1 - 2 | 306 | 103 | |
| AK120281.1 | - | 1 - 2 | 342 | 103 | ||
More Information
Set Membership: |
|

LOC9269259
Genomic Map