Assay Details
| Gene Symbol: LOC9269259 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777817.2 | XP_015633303.1 | 2 - 3 | 459 | 87 | |
| GenBank mRNA | AK059731.1 | - | 2 - 3 | 391 | 87 | |
| AK120281.1 | - | 2 - 3 | 427 | 87 | ||
More Information
Set Membership: |
|

LOC9269259
Genomic Map