Assay Details
| Gene Symbol: LOC9269259 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777817.2 | XP_015633303.1 | 3 - 4 | 524 | 75 | |
| GenBank mRNA | AK059731.1 | - | 3 - 4 | 456 | 75 | |
| AK120281.1 | - | 3 - 4 | 492 | 75 | ||
More Information
Set Membership: |
|

LOC9269259
Genomic Map