Assay Details
| Gene Symbol: LOC9269259 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777817.2 | XP_015633303.1 | 9 - 10 | 1154 | 86 | |
| GenBank mRNA | AK120281.1 | - | 8 - 9 | 3966 | 86 | |
More Information
Set Membership: |
|

LOC9269259
Genomic Map