Assay Details
| Gene Symbol: LOC4339683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK288455.1 | - | 1700 | 79 | ||
| Gene Symbol: LOC4333880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775647.2 | XP_015631133.1 | 5 - 6 | 1003 | 79 | |
| XM_015775648.2 | XP_015631134.1 | 5 - 6 | 947 | 79 | ||
| GenBank mRNA | AK062077.1 | - | 5 - 6 | 936 | 79 | |
| AK103065.1 | - | 5 - 6 | 933 | 79 | ||
| BT129313.1 | - | 4 - 5 | 453 | 79 | ||
| BT132195.1 | - | 3 - 4 | 279 | 79 | ||
| EU268000.1 | - | 5 - 6 | 912 | 79 | ||
| KC140113.1 | - | 5 - 6 | 973 | 79 | ||
More Information
Set Membership: |
|

LOC4339683
Genomic Map