Assay Details
Gene Symbol: LOC4327136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015767540.2 | XP_015623026.1 | 8 - 9 | 1880 | 78 | |
GenBank mRNA | AK066661.1 | - | 8 - 9 | 1878 | 78 | |
AK102967.1 | - | 4 - 5 | 612 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
