Assay Details
| Gene Symbol: LOC4327080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769464.2 | XP_015624950.1 | 1 - 2 | 244 | 107 | |
| GenBank mRNA | AK101188.1 | - | 1 - 2 | 237 | 107 | |
| AK101572.1 | - | 1 - 2 | 230 | 107 | ||
More Information
Set Membership: |
|

LOC4327080
Genomic Map