Assay Details
| Gene Symbol: LOC4327080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769464.2 | XP_015624950.1 | 6 - 7 | 1484 | 112 | |
| GenBank mRNA | AK067829.1 | - | 5 - 6 | 1907 | 112 | |
| AK101188.1 | - | 6 - 7 | 1477 | 112 | ||
| AK101572.1 | - | 6 - 7 | 1470 | 112 | ||
More Information
Set Membership: |
|

LOC4327080
Genomic Map