Assay Details
| Gene Symbol: LOC4327192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770383.2 | XP_015625869.1 | 11 - 12 | 3362 | 65 | |
| GenBank mRNA | AJ495800.1 | - | 2 - 3 | 267 | 65 | |
More Information
Set Membership: |
|

LOC4327192
Genomic Map