Assay Details
| Gene Symbol: LOC4326266 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761488.2 | XP_015616974.1 | 5 - 6 | 1027 | 87 | |
| GenBank mRNA | AK068875.1 | - | 5 - 6 | 1027 | 87 | |
More Information
Set Membership: |
|

LOC4326266
Genomic Map